Assay Details
Target Gene Details
Entrez Gene ID: | 13070 |
Gene Name: | cytochrome P450, family 11, subfamily a, polypeptide 1 |
Gene Aliases: |
Cyp11a, Cypxia1, D9Ertd411e, P450scc, Scc, cscc |
Location: |
Chr.9:58015017-58027023 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp11a1 | NM_019779.3 | 3 | 575 | NP_062753.3 |
| AF195119.1 | 3 | 575 | AAF03897.1 | |
| AK077945.1 | 3 | 596 | ||
| BC068264.1 | 3 | 594 | AAH68264.1 |

Genomic Map