Assay Details
Target Gene Details
Entrez Gene ID: | 68318 |
Gene Name: | anterior pharynx defective 1c homolog (C. elegans) |
Gene Aliases: |
0610008A10Rik, APH-1b, Aph1b |
Location: |
Chr.9:66814994-66834779 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aph1c | NM_026674.3 | NP_080950.1 | ||
| AB197123.1 | BAE02661.1 | |||
| AK002310.1 | BAB22004.1 | |||
| AK155147.1 | 1 | 42 | BAE33077.1 | |
| BC050923.1 | AAH50923.1 | |||
| BC063254.1 | AAH63254.1 |
More Information
Set Membership: |
|

Genomic Map