Assay Details
Target Gene Details
Entrez Gene ID: | 382077 |
Gene Name: | coiled-coil domain containing 33 |
Gene Aliases: |
4930535E21Rik, Gm1117, Tzip1 |
Location: |
Chr.9:58028673-58118823 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc33 | NM_001166282.1 | NP_001159754.1 | ||
| NM_029212.3 | NP_083488.1 | |||
| AB048860.1 | BAD08245.1 | |||
| AK145264.1 | BAE26332.1 | |||
| BC116291.1 | ||||
| BC116292.1 |
Target Gene Details
Entrez Gene ID: | 330951 |
Gene Name: | predicted gene 16130 |
Gene Aliases: |
E330033L03, OTTMUSG00000030814 |
Location: |
Chr.9:58032274-58048997 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16130 | NM_001243265.1 | 2 | 814 | NP_001230194.1 |
| AK054504.1 | 2 | 815 |

Genomic Map