Assay Details
Target Gene Details
Entrez Gene ID: | 225215 |
Gene Name: | ribosomal L24 domain containing 1 |
Gene Aliases: |
2410159K22Rik |
Location: |
Chr.9:73113469-73123333 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rsl24d1 | NM_198609.2 | NP_941011.1 | ||
| AK153492.1 | BAE32040.1 | |||
| AK160194.1 | BAE35686.1 | |||
| AK164856.1 | BAE37944.1 | |||
| BC003885.1 | AAH03885.1 | |||
| BC089481.1 | AAH89481.1 |
More Information
Set Membership: |
|

Genomic Map