Assay Details
Target Gene Details
Entrez Gene ID: | 235044 |
Gene Name: | lipid phosphate phosphatase-related protein type 2 |
Gene Aliases: |
PRG-4 |
Location: |
Chr.9:21936422-21948907 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lppr2 | NM_001290299.1 | NP_001277228.1 | ||
| NM_144935.1 | NP_659184.1 | |||
| AK082453.1 | BAC38494.1 | |||
| AY438023.1 | AAR98841.1 | |||
| BC018242.1 | AAH18242.1 | |||
| BC023082.1 |
More Information
Set Membership: |
|

Genomic Map