Assay Details
Target Gene Details
Entrez Gene ID: | 235415 |
Gene Name: | complexin 3 |
Gene Aliases: |
BC016632, CpxIII, ERGIC-53L, ERGL, Lamn1l, Lman1l |
Location: |
Chr.9:57599992-57606281 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cplx3 | NM_146223.3 | NP_666335.1 | ||
| AK044211.1 | BAC31818.1 | |||
| AY264290.1 | AAP22133.1 | |||
| BC016632.1 | AAH16632.1 | |||
| BC024854.1 | AAH24854.1 | |||
| BC033343.1 | AAH33343.1 |

Genomic Map