Assay Details
Target Gene Details
Entrez Gene ID: | 112418 |
Gene Name: | RIKEN cDNA 1700102P08 gene |
Gene Aliases: |
AI593206 |
Location: |
Chr.9:108392834-108410966 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700102P08Rik |
Target Gene Details
Entrez Gene ID: | 434438 |
Gene Name: | coiled-coil domain containing 36 |
Gene Aliases: |
- |
Location: |
Chr.9:108403502-108431841 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc36 | NM_001135198.1 | 6 | 600 | NP_001128670.1 |
| BC058624.1 | 6 | 605 | AAH58624.1 |

Genomic Map