Assay Details
Target Gene Details
Entrez Gene ID: | 78025 |
Gene Name: | RIKEN cDNA 4930540M03 gene |
Gene Aliases: |
- |
Location: |
Chr.9:15619857-15641220 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930540M03Rik | NR_040746.1 | |||
| AK019729.1 |
Target Gene Details
Entrez Gene ID: | 234964 |
Gene Name: | coiled-coil domain containing 67 |
Gene Aliases: |
4933401K09Rik, Deup1 |
Location: |
Chr.9:15559864-15628338 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc67 | NM_181816.2 | NP_861537.2 | ||
| AK030153.1 | ||||
| BY069891.1 |
More Information
Set Membership: |
|

Genomic Map