Assay Details
Target Gene Details
Entrez Gene ID: | 234964 |
Gene Name: | coiled-coil domain containing 67 |
Gene Aliases: |
4933401K09Rik, Deup1 |
Location: |
Chr.9:15559864-15628338 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc67 | NM_181816.2 | 3 | 268 | NP_861537.2 |
| AK030153.1 | 3 | 251 | ||
| BC096547.1 | 2 | 161 | AAH96547.1 | |
| BY069891.1 | 3 | 269 | ||
| KC211186.1 | 2 | 112 |
More Information
Set Membership: |
|

Genomic Map