Assay Details
Target Gene Details
Entrez Gene ID: | 234964 |
Gene Name: | coiled-coil domain containing 67 |
Gene Aliases: |
4933401K09Rik, Deup1 |
Location: |
Chr.9:15559864-15628338 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc67 | NM_181816.2 | 7 | 780 | NP_861537.2 |
| AK030153.1 | 7 | 763 | ||
| AK077147.1 | 2 | 158 | BAC36643.1 | |
| BC096547.1 | 6 | 673 | AAH96547.1 | |
| KC211186.1 | 6 | 624 |
More Information
Set Membership: |
|

Genomic Map