Assay Details
Target Gene Details
Entrez Gene ID: | 21922 |
Gene Name: | C-type lectin domain family 3, member b |
Gene Aliases: |
Tna |
Location: |
Chr.9:123150946-123157432 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec3b | NM_011606.2 | NP_035736.2 | ||
| BC035043.1 | AAH35043.1 | |||
| BY114395.1 | ||||
| U08595.1 | AAA96811.1 | |||
| X79199.1 | CAA55791.1 |
More Information
Set Membership: |
|

Genomic Map