Assay Details
Target Gene Details
Entrez Gene ID: | 76524 |
Gene Name: | ceroid-lipofuscinosis, neuronal 6 |
Gene Aliases: |
1810065L06Rik, AW743417, D9Bwg1455e, nclf |
Location: |
Chr.9:62838787-62852002 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cln6 | NM_001033175.2 | 3 | 323 | NP_001028347.1 |
| AK154414.1 | 3 | 324 | BAE32568.1 | |
| AK154551.1 | 3 | 314 | ||
| BC116794.1 | 2 | 149 | ||
| BC116798.1 | 2 | 149 |

Genomic Map