Assay Details
Target Gene Details
Entrez Gene ID: | 11481 |
Gene Name: | activin receptor IIB |
Gene Aliases: |
ActRIIB |
Location: |
Chr.9:119401583-119442134 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acvr2b | NM_007397.2 | 5 | 853 | NP_031423.1 |
| AK137432.1 | 5 | 604 | ||
| BC106189.1 | 6 | 838 | AAI06190.1 | |
| M84120.1 | 5 | 851 | AAA37172.1 |
More Information
Set Membership: |
|

Genomic Map