Assay Details
Target Gene Details
Entrez Gene ID: | 12442 |
Gene Name: | cyclin B2 |
Gene Aliases: |
CycB2 |
Location: |
Chr.9:70407689-70421554 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnb2 | NM_007630.2 | NP_031656.2 | ||
| AK013312.1 | BAB28785.1 | |||
| AK076122.1 | BAC36200.1 | |||
| AK177442.1 | ||||
| BC008247.1 | AAH08247.1 | |||
| CT010286.1 | CAJ18494.1 | |||
| X66032.1 | CAA46831.1 |
More Information
Set Membership: |
|

Genomic Map