Assay Details
Target Gene Details
Entrez Gene ID: | 12772 |
Gene Name: | chemokine (C-C motif) receptor 2 |
Gene Aliases: |
Cc-ckr-2, Ccr2a, Ccr2b, Ckr2, Ckr2a, Ckr2b, Cmkbr2, mJe-r |
Location: |
Chr.9:124101918-124109140 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccr2 | NM_009915.2 | NP_034045.1 | ||
| AK154590.1 | 1 | 79 | BAE32695.1 | |
| U56819.1 | AAC52784.1 |

Genomic Map