Assay Details
Target Gene Details
Entrez Gene ID: | 109652 |
Gene Name: | aminoacylase 1 |
Gene Aliases: |
1110014J22Rik, Acy-1 |
Location: |
Chr.9:106432981-106438236 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acy1 | NM_001276442.1 | NP_001263371.1 | ||
| NM_025371.3 | NP_079647.1 | |||
| AK002423.1 | BAB22090.1 | |||
| AK003703.1 | BAB22948.1 | |||
| BC005631.1 | AAH05631.1 | |||
| BX529990.1 |

Genomic Map