Assay Details
Target Gene Details
Entrez Gene ID: | 109652 |
Gene Name: | aminoacylase 1 |
Gene Aliases: |
1110014J22Rik, Acy-1 |
Location: |
Chr.9:106432981-106438236 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acy1 | NM_001276442.1 | 15 | 1301 | NP_001263371.1 |
| NM_025371.3 | 15 | 1305 | NP_079647.1 | |
| AK002423.1 | 15 | 1291 | BAB22090.1 | |
| AK003703.1 | 15 | 1306 | BAB22948.1 | |
| BC005631.1 | 15 | 1309 | AAH05631.1 | |
| BX529990.1 | 3 | 269 |

Genomic Map