Assay Details
Target Gene Details
Entrez Gene ID: | 244867 |
Gene Name: | Rho GTPase activating protein 20 |
Gene Aliases: |
6530403F17Rik, A530023E23Rik, RARhoGAP, mKIAA1391 |
Location: |
Chr.9:51765087-51853856 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap20 | NM_175535.3 | 13 | 1715 | NP_780744.2 |
| AK129345.1 | 10 | 1204 | BAC98155.1 |
Target Gene Details
Entrez Gene ID: | 408028 |
Gene Name: | Rho GTPase activating protein 20, opposite strand |
Gene Aliases: |
9230115E21Rik |
Location: |
Chr.9:51839493-51875885 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap20os | NR_033560.1 | |||
| AK033819.1 | ||||
| AK165145.1 | BAE38048.1 |
More Information
Set Membership: |
|

Genomic Map