Assay Details
Target Gene Details
Entrez Gene ID: | 214763 |
Gene Name: | Mab-21 domain containing 1 |
Gene Aliases: |
E330016A19Rik, cGAS |
Location: |
Chr.9:78430518-78443524 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mb21d1 | NM_173386.5 | 3 | 1157 | NP_775562.2 |
| NR_125721.1 | 3 | 1201 | ||
| AK030292.1 | 3 | 1158 | BAC26881.1 | |
| AK054330.1 | 3 | 1091 | BAC35733.1 | |
| AK078662.1 | 3 | 1068 | BAC37351.1 | |
| AK145268.1 | 3 | 1097 | BAE26335.1 | |
| AK154165.1 | 3 | 1126 | ||
| BC044854.1 | 3 | 861 | ||
| BC052196.1 | 3 | 1067 | ||
| BC145651.1 | 3 | 1127 | ||
| BC145653.1 | 3 | 1127 | ||
| KC294567.1 | 3 | 998 |

Genomic Map