Assay Details
Target Gene Details
Entrez Gene ID: | 68283 |
Gene Name: | RIKEN cDNA 9530077C05 gene |
Gene Aliases: |
1110003N12Rik, Kiaa0895, mKIAA0895 |
Location: |
Chr.9:22387732-22444684 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9530077C05Rik | NM_026739.1 | NP_081015.1 | ||
| AK021129.1 | 1 | 38 | BAC25646.1 | |
| AK160343.1 | 1 | 51 | ||
| AK170604.1 | 1 | 33 | ||
| AK220359.1 | 1 | 66 | BAD90420.1 | |
| BC054761.1 | AAH54761.2 |

Genomic Map