Assay Details
Target Gene Details
Entrez Gene ID: | 68283 |
Gene Name: | RIKEN cDNA 9530077C05 gene |
Gene Aliases: |
1110003N12Rik, Kiaa0895, mKIAA0895 |
Location: |
Chr.9:22387732-22444684 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9530077C05Rik | NM_026739.1 | NP_081015.1 | ||
| AK003384.1 | BAB22754.1 | |||
| AK170604.1 | ||||
| AK220359.1 | BAD90420.1 | |||
| BC039998.1 | AAH39998.1 | |||
| BC054761.1 | AAH54761.2 |

Genomic Map