Assay Details
Target Gene Details
Entrez Gene ID: | 434437 |
Gene Name: | aminomethyltransferase |
Gene Aliases: |
EG434437 |
Location: |
Chr.9:108296922-108301722 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amt | NM_001013814.1 | 8 | 965 | NP_001013836.1 |
| BC132277.1 | 8 | 1017 | ||
| BC132279.1 | 8 | 1017 |
More Information
Set Membership: |
|

Genomic Map