Assay Details
Target Gene Details
Entrez Gene ID: | 70726 |
Gene Name: | angiopoietin-like 6 |
Gene Aliases: |
6330404E11Rik, AGF, ARP3, Arp5 |
Location: |
Chr.9:20873808-20879816 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Angptl6 | NM_145154.2 | NP_660136.1 | ||
| AB054065.1 | BAB91249.1 | |||
| BC025904.1 | AAH25904.1 |
More Information
Set Membership: |
|

Genomic Map