Assay Details
Target Gene Details
Entrez Gene ID: | 170752 |
Gene Name: | beta-carotene oxygenase 2 |
Gene Aliases: |
B-diox-II, Bcdo2, Bcmo2, CMO2 |
Location: |
Chr.9:50533087-50555220 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bco2 | NM_133217.3 | NP_573480.1 | ||
| BB624957.1 |
Target Gene Details
Entrez Gene ID: | 16173 |
Gene Name: | interleukin 18 |
Gene Aliases: |
Igif, Il-18 |
Location: |
Chr.9:50554705-50581841 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il18 | AK078100.1 |
More Information
Set Membership: |
|

Genomic Map