Assay Details
Target Gene Details
Entrez Gene ID: | 319278 |
Gene Name: | RIKEN cDNA A230050P20 gene |
Gene Aliases: |
- |
Location: |
Chr.9:20868592-20874307 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A230050P20Rik | NM_175687.2 | NP_783618.1 | ||
| AK034516.1 | ||||
| AK149161.1 | BAE28753.1 | |||
| AK157146.1 | BAE33979.1 | |||
| BC031510.1 | ||||
| BC043448.1 | AAH43448.1 |
More Information
Set Membership: |
|

Genomic Map