Assay Details
Target Gene Details
Entrez Gene ID: | 12796 |
Gene Name: | cathelicidin antimicrobial peptide |
Gene Aliases: |
CAP18, Cnlp, Cramp, FALL39, MCLP |
Location: |
Chr.9:109847377-109849456 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camp | NM_009921.2 | NP_034051.2 | ||
| AK158005.1 | BAE34312.1 | |||
| BC120752.1 | ||||
| BC120754.1 | ||||
| BY582734.1 | ||||
| U43409.1 | AAA85898.1 | |||
| X94353.1 | CAA64078.1 |
More Information
Set Membership: |
|

Genomic Map