Assay Details
Target Gene Details
Entrez Gene ID: | 12500 |
Gene Name: | CD3 antigen, delta polypeptide |
Gene Aliases: |
T3d |
Location: |
Chr.9:44981769-44987052 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd3d | NM_013487.3 | 3 | 440 | NP_038515.3 |
| AK042172.1 | 3 | 442 | BAE20624.1 | |
| AK050629.1 | 3 | 429 | BAE20683.1 | |
| AK153843.1 | 3 | 405 | BAE32208.1 | |
| AK154064.1 | 3 | 402 | BAE32350.1 | |
| BC062807.1 | 3 | 345 | AAH62807.1 | |
| EF382667.1 | 3 | 340 | ||
| HM536173.1 | 3 | 340 | ||
| X02339.1 | 3 | 308 | CAA26198.1 |

Genomic Map