Assay Details
Target Gene Details
Entrez Gene ID: | 108153 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 7 |
Gene Aliases: |
ADAM-TS7, ADAMTS7B |
Location: |
Chr.9:90162978-90200102 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamts7 | NM_001003911.2 | 8 | 1465 | NP_001003911.2 |
| AY551090.1 | 8 | 1198 | AAT36307.1 | |
| BC058991.1 | 5 | 443 | AAH58991.1 | |
| BC141173.1 | 8 | 1241 | ||
| BC145491.1 | 8 | 1241 |
More Information
Set Membership: |
|

Genomic Map