Assay Details
Target Gene Details
Entrez Gene ID: | 68283 |
Gene Name: | RIKEN cDNA 9530077C05 gene |
Gene Aliases: |
1110003N12Rik, Kiaa0895, mKIAA0895 |
Location: |
Chr.9:22387732-22444684 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9530077C05Rik |
Target Gene Details
Entrez Gene ID: | 68743 |
Gene Name: | anillin, actin binding protein |
Gene Aliases: |
1110037A17Rik, 2900037I21Rik, C78101, Scraps |
Location: |
Chr.9:22331214-22389696 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anln | NM_028390.3 | 1 | 167 | NP_082666.2 |
| BC027141.1 | 1 | 131 | ||
| BC032164.1 | 1 | 185 | AAH32164.1 | |
| CN458410.1 | 1 | 84 |
More Information
Set Membership: |
|

Genomic Map