Assay Details
Target Gene Details
Entrez Gene ID: | 12502 |
Gene Name: | CD3 antigen, gamma polypeptide |
Gene Aliases: |
Ctg-3, Ctg3, T3g |
Location: |
Chr.9:44969572-44980431 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd3g | NM_009850.2 | NP_033980.1 | ||
| AK133993.1 | 1 | 1864 | ||
| AK134049.1 | BAE21993.1 | |||
| AK153980.1 | BAE32293.1 | |||
| AK153987.1 | BAE32297.1 | |||
| BC027528.1 | AAH27528.1 | |||
| EF474453.1 | ||||
| GU456630.1 | ||||
| M58149.1 | AAA63289.1 | |||
| Y00635.1 | CAA68667.1 |

Genomic Map