Assay Details
Target Gene Details
Entrez Gene ID: | 12530 |
Gene Name: | cell division cycle 25A |
Gene Aliases: |
D9Ertd393e |
Location: |
Chr.9:109875337-109893895 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc25a | NM_007658.3 | NP_031684.3 | ||
| AK036499.1 | ||||
| AK077609.1 | ||||
| AK077991.1 | ||||
| AK142334.1 | BAE25035.1 | |||
| BC046296.1 | AAH46296.1 | |||
| U27323.1 | AAA85580.1 |
Target Gene Details
Entrez Gene ID: | 102465660 |
Gene Name: | microRNA 7089 |
Gene Aliases: |
Gm27418, mmu-mir-7089 |
Location: |
Chr.9:109889643-109889710 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir7089 | NR_106057.1 |
More Information
Set Membership: |
|

Genomic Map