Assay Details
Target Gene Details
Entrez Gene ID: | 12539 |
Gene Name: | cell division cycle 37 |
Gene Aliases: |
p50, p50Cdc37 |
Location: |
Chr.9:21138541-21149977 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc37 | NM_016742.4 | NP_058022.1 | ||
| AK010494.1 | BAB26984.1 | |||
| AK013255.1 | BAB28749.1 | |||
| AK145671.1 | BAE26580.1 | |||
| AK168651.1 | BAE40508.1 | |||
| AK206081.1 | 1 | 132 | ||
| AK213273.1 | 2 | 300 | ||
| BC060079.1 | AAH60079.1 | |||
| U43076.1 | AAB18761.1 |
More Information
Set Membership: |
|

Genomic Map