Assay Details
Target Gene Details
Entrez Gene ID: | 12836 |
Gene Name: | collagen, type VII, alpha 1 |
Gene Aliases: |
AW209154 |
Location: |
Chr.9:108953575-108984875 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 61 - Exon 62 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col7a1 | NM_007738.3 | NP_031764.2 | ||
| BC061507.1 | AAH61507.1 | |||
| CJ045811.1 | ||||
| S63654.1 | AAB27492.1 | |||
| U32107.1 | AAB66593.1 |
Target Gene Details
Entrez Gene ID: | 751536 |
Gene Name: | microRNA 711 |
Gene Aliases: |
Mirn711, mir-711, mmu-mir-711 |
Location: |
Chr.9:108969922-108970003 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir711 | NR_030492.1 |
More Information
Set Membership: |
|

Genomic Map