Assay Details
Target Gene Details
Entrez Gene ID: | 68644 |
Gene Name: | abhydrolase domain containing 14A |
Gene Aliases: |
1110013B16Rik, AW558221, Dorz1 |
Location: |
Chr.9:106440051-106451384 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd14a |
Target Gene Details
Entrez Gene ID: | 76491 |
Gene Name: | abhydrolase domain containing 14b |
Gene Aliases: |
1810013B01Rik |
Location: |
Chr.9:106447737-106452923 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd14b | NM_029631.3 | NP_083907.3 | ||
| AK007469.1 | ||||
| BC019410.1 | AAH19410.1 | |||
| CJ186593.1 |

Genomic Map