Assay Details
Target Gene Details
Entrez Gene ID: | 12362 |
Gene Name: | caspase 1 |
Gene Aliases: |
ICE, Il1bc |
Location: |
Chr.9:5298517-5307281 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Casp1 | NM_009807.2 | NP_033937.2 | ||
| AK132826.1 | ||||
| AK163069.1 | 3 | 1779 | ||
| BC008152.1 | AAH08152.1 | |||
| L03799.1 | AAA39306.1 | |||
| L28095.1 | AAA20209.1 |
More Information
Set Membership: |
|

Genomic Map