Assay Details
Target Gene Details
Entrez Gene ID: | 66070 |
Gene Name: | CWC15 homolog (S. cerevisiae) |
Gene Aliases: |
0610040D20Rik, 2900052N06Rik, Ed1, c11orf5, mED1 |
Location: |
Chr.9:14500606-14510620 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cwc15 | NM_023153.3 | NP_075642.1 | ||
| AJ250394.1 | CAB96547.1 | |||
| AK002864.1 | BAB22414.1 | |||
| AK003202.1 | BAB22638.1 | |||
| AK003591.1 | BAB22880.1 | |||
| AK003834.2 | BAB23026.1 | |||
| AK145249.1 | BAE26327.1 | |||
| AK167776.1 | BAE39808.1 | |||
| AK168365.1 | BAE40300.1 | |||
| AY842452.1 | AAW32096.1 | |||
| BC004726.1 | AAH04726.1 | |||
| BQ840384.1 |

Genomic Map