Assay Details
Target Gene Details
Entrez Gene ID: | 59046 |
Gene Name: | cAMP-regulated phosphoprotein 19 |
Gene Aliases: |
19kDa, 2700024H10Rik, ARPP-19, AW559096, Arpp12 |
Location: |
Chr.9:75037614-75060313 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arpp19 | NM_001142655.1 | NP_001136127.1 |

Genomic Map