Assay Details
Target Gene Details
Entrez Gene ID: | 11768 |
Gene Name: | adaptor protein complex AP-1, mu 2 subunit |
Gene Aliases: |
D9Ertd818e, [m]1B, mu1B |
Location: |
Chr.9:21294613-21312349 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap1m2 | NM_001110300.1 | NP_001103770.1 | ||
| NM_009678.2 | NP_033808.2 | |||
| AF067146.1 | AAD28085.1 | |||
| AK010478.1 | BAB26971.1 | |||
| AK160161.1 | ||||
| BC003704.1 | AAH03704.1 |
More Information
Set Membership: |
|

Genomic Map