Assay Details
Target Gene Details
Entrez Gene ID: | 23918 |
Gene Name: | inosine 5'-phosphate dehydrogenase 2 |
Gene Aliases: |
IMPD |
Location: |
Chr.9:108560291-108565584 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Impdh2 | NM_011830.3 | NP_035960.2 | ||
| AK002676.1 | ||||
| AK011777.1 | ||||
| AK143505.1 | BAE25403.1 | |||
| AK146362.1 | ||||
| AK150713.1 | BAE29791.1 | |||
| AK151233.1 | BAE30225.1 | |||
| AK151706.1 | BAE30628.1 | |||
| AK152691.1 | BAE31422.1 | |||
| AK159822.1 | BAE35402.1 | |||
| AK160569.1 | BAE35879.1 | |||
| AK166667.1 | BAE38929.1 | |||
| AK169129.1 | BAE40908.1 | |||
| AK169788.1 | BAE41367.1 | |||
| AK215538.1 | ||||
| BC010314.1 | AAH10314.1 | |||
| BC052671.1 | AAH52671.1 | |||
| M33934.1 | AAA39311.1 | |||
| M98333.1 | AAA20181.1 |
More Information
Set Membership: |
|

Genomic Map