Assay Details
Target Gene Details
Entrez Gene ID: | 17384 |
Gene Name: | matrix metallopeptidase 10 |
Gene Aliases: |
AV377895, MMP-10, SL-2 |
Location: |
Chr.9:7502342-7510242 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmp10 | NM_019471.3 | NP_062344.1 | ||
| AK020292.1 | BAB32058.1 | |||
| AY622970.1 | AAT46401.1 | |||
| BC130027.1 | ||||
| X76537.1 | ||||
| Y13185.1 | CAA73641.1 |
More Information
Set Membership: |
|

Genomic Map