Assay Details
Target Gene Details
Entrez Gene ID: | 67834 |
Gene Name: | isocitrate dehydrogenase 3 (NAD+) alpha |
Gene Aliases: |
1110003P10Rik, 1500012E04Rik, AA407078, AI316514 |
Location: |
Chr.9:54586511-54604662 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Idh3a | NM_029573.2 | NP_083849.1 | ||
| AK010065.1 | BAB26679.1 | |||
| AK032787.1 | BAC28021.1 | |||
| AK047951.1 | BAC33199.1 | |||
| AK084523.1 | ||||
| AK117103.1 | ||||
| AK150618.1 | BAE29708.1 | |||
| AK151304.1 | BAE30286.1 | |||
| AK152353.1 | BAE31145.1 | |||
| AK153459.1 | BAE32011.1 | |||
| AK158646.1 | BAE34596.1 | |||
| AK159051.1 | BAE34785.1 | |||
| AK159666.1 | ||||
| AK160054.1 | ||||
| AK164038.1 | ||||
| AK167822.1 | ||||
| AK168034.1 | ||||
| AK168049.1 | BAE40031.1 | |||
| AK168149.1 | BAE40114.1 | |||
| AK169152.1 | BAE40931.1 | |||
| BC034273.1 | AAH34273.1 | |||
| BC049956.1 | AAH49956.1 |
More Information
Set Membership: |
|

Genomic Map