Assay Details
Target Gene Details
Entrez Gene ID: | 67320 |
Gene Name: | IQ motif containing F4 |
Gene Aliases: |
1700042N06Rik, mtIQ1 |
Location: |
Chr.9:106568319-106570967 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqcf4 | NM_026090.2 | 2 | 235 | NP_080366.2 |
| AK006688.1 | 2 | 234 | BAB24704.1 | |
| AK007079.1 | ||||
| AK018930.1 | ||||
| BC048652.1 | 2 | 240 | ||
| BC061014.1 | 2 | 235 | AAH61014.1 | |
| DQ153246.1 | 2 | 264 | AAZ78214.1 |

Genomic Map