Assay Details
Target Gene Details
Entrez Gene ID: | 68165 |
Gene Name: | ferredoxin 1-like |
Gene Aliases: |
B230118G17Rik |
Location: |
Chr.9:21067520-21073514 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fdx1l | NM_001039824.2 | NP_001034913.1 | ||
| AK008401.2 | BAB25650.1 | |||
| AK010211.1 | BAB26771.1 | |||
| AK020979.1 | BAB32267.1 | |||
| BC061189.1 | AAH61189.1 |
Target Gene Details
Entrez Gene ID: | 100568458 |
Gene Name: | predicted gene, 38431 |
Gene Aliases: |
Raver1-fdx1l |
Location: |
Chr.9:21067514-21092008 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm38431 | NR_038081.2 | |||
| AK004919.1 | BAB23670.1 | |||
| AY275472.1 | AAP33691.1 |
More Information
Set Membership: |
|

Genomic Map