Assay Details
Target Gene Details
Entrez Gene ID: | 235416 |
Gene Name: | lectin, mannose-binding 1 like |
Gene Aliases: |
BC020188, slamp |
Location: |
Chr.9:57606717-57621419 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lman1l | NM_199222.3 | NP_954692.2 | ||
| AB211986.1 | BAE16262.1 |
More Information
Set Membership: |
|

Genomic Map