Assay Details
Target Gene Details
Entrez Gene ID: | 72454 |
Gene Name: | coiled-coil domain containing 71 |
Gene Aliases: |
2600016J21Rik |
Location: |
Chr.9:108452571-108465945 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc71 | NM_133744.4 | NP_598505.1 | ||
| AK011229.1 | ||||
| AK044601.1 | BAC31997.1 | |||
| AK079333.1 | ||||
| AK085544.1 | ||||
| AK086560.1 | BAC39691.1 | |||
| AK136982.1 | ||||
| AK149878.1 | BAE29140.1 | |||
| AK161396.1 | 1 | 88 | BAE36372.1 | |
| BC018518.1 | AAH18518.1 | |||
| CJ146452.1 |
Target Gene Details
Entrez Gene ID: | 78267 |
Gene Name: | kelch domain containing 8B |
Gene Aliases: |
4931406O17Rik |
Location: |
Chr.9:108447636-108461581 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhdc8b | NM_030075.2 | 1 | 76 | NP_084351.1 |
| AK019849.1 | 1 | 76 | BAB31879.1 |
More Information
Set Membership: |
|

Genomic Map