Assay Details
Target Gene Details
Entrez Gene ID: | 78267 |
Gene Name: | kelch domain containing 8B |
Gene Aliases: |
4931406O17Rik |
Location: |
Chr.9:108447636-108461581 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhdc8b | NM_030075.2 | NP_084351.1 | ||
| AK019849.1 | BAB31879.1 | |||
| AK154428.1 | BAE32579.1 | |||
| BC048918.1 | AAH48918.1 |

Genomic Map