Assay Details
Target Gene Details
Entrez Gene ID: | 78267 |
Gene Name: | kelch domain containing 8B |
Gene Aliases: |
4931406O17Rik |
Location: |
Chr.9:108447636-108461581 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhdc8b | NM_030075.2 | 9 | 1897 | NP_084351.1 |
| AK019849.1 | 9 | 1895 | BAB31879.1 | |
| AK154428.1 | 9 | 1998 | BAE32579.1 | |
| AK215637.1 | ||||
| BC048918.1 | 4 | 860 | AAH48918.1 |

Genomic Map