Assay Details
Target Gene Details
Entrez Gene ID: | 234911 |
Gene Name: | matrix metallopeptidase 27 |
Gene Aliases: |
Gm180 |
Location: |
Chr.9:7571396-7581885 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmp27 | NM_001030289.1 | 1 | 76 | NP_001025460.1 |
| AK155813.1 | 1 | 141 | ||
| AK159182.1 | 1 | 72 |
More Information
Set Membership: |
|

Genomic Map