Assay Details
Target Gene Details
Entrez Gene ID: | 64931 |
Gene Name: | IZUMO1 receptor, JUNO |
Gene Aliases: |
0910001L11Rik, C86255, Folbp3, Folr4, Juno |
Location: |
Chr.9:14885814-14903951 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Izumo1r | NM_022888.2 | NP_075026.1 | ||
| NM_176807.4 | NP_789777.1 | |||
| AF250145.1 | AAG36877.1 | |||
| AK003087.1 | ||||
| AK041596.1 | BAC30998.1 | |||
| AK083306.1 | BAC38857.1 | |||
| BC028431.1 | AAH28431.1 | |||
| EU326437.1 | ||||
| EU326438.1 | ||||
| EU326439.1 | ||||
| EU326440.1 | ||||
| EU326441.1 | ||||
| EU326442.1 |

Genomic Map