Assay Details
Target Gene Details
Entrez Gene ID: | 68121 |
Gene Name: | centrosomal protein 70 |
Gene Aliases: |
6720484E09Rik, Bite, C030018L16Rik |
Location: |
Chr.9:99183595-99302065 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep70 |
Target Gene Details
Entrez Gene ID: | 623459 |
Gene Name: | predicted gene 6432 |
Gene Aliases: |
EG623459 |
Location: |
Chr.9:99229376-99237239 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm6432 | NM_001244762.1 | NP_001231691.1 | ||
| BU939370.1 |

Genomic Map